Heptadecylcyclohexane ≥97.0%

Supplier: TCI America

Synonyms: 1-Cyclohexylheptadecane

H0443-5G
TCH0443-005GEA 149 USD
TCH0443-005G
Heptadecylcyclohexane ≥97.0%
Heptadecylcyclohexane

CAS Number: 19781-73-8
MDL Number: MFCD00051453
Molecular Formula: C23H46
Molecular Weight: 322.62
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 397
Melting point (°C): 36
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Cyclohexylheptadecane

Formula: C₂₃H₄₆
MW: 322.62 g/mol
Boiling Pt: 397 °C
Melting Pt: 36 °C
Density: 0.8332 g/cm³ (18 °C)
MDL Number: MFCD00051453
CAS Number: 19781-73-8
EINECS: 243-308-2
https://us.vwr.com/stibo/web/std.lang.all/04/56/28970456.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR