Heptylcyclohexane ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1-Cyclohexylheptane

H0442-25ML
TCH0442-025MLEA 195.2 USD
TCH0442-025ML
Heptylcyclohexane ≥99.0%
Heptylcyclohexane

CAS Number: 5617-41-4
MDL Number: MFCD00019404
Molecular Formula: C13H26
Molecular Weight: 182.35
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 244
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: 1-Cyclohexylheptane

Formula: C₁₃H₂₆
MW: 182.35 g/mol
Boiling Pt: 244 °C
Melting Pt: –30 °C
Density: 0.81 (20 °C)
MDL Number: MFCD00019404
CAS Number: 5617-41-4
EINECS: 227-041-9
https://us.vwr.com/stibo/web/std.lang.all/04/55/28970455.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR