8-Hexadecyne ≥99.0%

Supplier: TCI America

H0434-5ML
TCH0434-005MLEA 227 USD
TCH0434-005ML
8-Hexadecyne ≥99.0%
8-Hexadecyne

CAS Number: 19781-86-3
MDL Number: MFCD00041664
Molecular Formula: C16H30
Molecular Weight: 222.42
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.80

Formula: C₁₆H₃₀
MW: 222.41 g/mol
Density: 0.80 (20 °C)
MDL Number: MFCD00041664
CAS Number: 19781-86-3

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR