1-Heptadecene ≥90.0%

Supplier: TCI America

H0372-25ML
TCH0372-025MLEA 124.85 USD
TCH0372-025ML
1-Heptadecene ≥90.0%
1-Heptadecene

CAS Number: 6765-39-5
MDL Number: MFCD00009000
Molecular Formula: C17H34
Molecular Weight: 238.46
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 300
Flash Point (°C): 110
Freezing point (°C): 10
Specific Gravity (20/20): 0.79

Formula: C₁₇H₃₄
MW: 238.46 g/mol
Boiling Pt: 300 °C
Melting Pt: 10 °C
Density: 0.79 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009000
CAS Number: 6765-39-5
EINECS: 229-825-6
https://us.vwr.com/stibo/web/std.lang.all/04/03/28970403.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR