2-Hydroxy-9-fluorenone ≥90.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Hydroxy-9H-fluoren-9-one
H0329-1G
TCH0329-001GEA
151
USD
TCH0329-001G
2-Hydroxy-9-fluorenone ≥90.0% (by HPLC, titration analysis)
2-Hydroxy-9-fluorenone
CAS Number: 6949-73-1
MDL Number: MFCD00001155
Molecular Formula: C13H8O2
Molecular Weight: 196.21
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 205
Formula:
C₁₃H₈O₂ MW: 196.21 g/mol Boiling Pt: 353 °C (1013 hPa) Melting Pt: 205 °C |
MDL Number:
MFCD00001155 CAS Number: 6949-73-1 EINECS: 230-119-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...