Hexylcyclohexane ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Cyclohexylhexane
H0137-10ML
H0137-25ML
TCH0137-25MLEA
281.08
USD
TCH0137-25ML
TCH0137-010ML
Hexylcyclohexane ≥98.0%
Hexylcyclohexane
CAS Number: 4292-75-5
MDL Number: MFCD00019403
Molecular Formula: C12H24
Molecular Weight: 168.32
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 221
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: 1-Cyclohexylhexane
Formula:
C₁₂H₂₄ MW: 168.32 g/mol Boiling Pt: 221 °C Melting Pt: –41 °C Density: 0.81 (20 °C) |
MDL Number:
MFCD00019403 CAS Number: 4292-75-5 EINECS: 224-294-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...