Isobutyl hexanoate ≥98.0%

Supplier: TCI America

Synonyms: Hexanoic acid Isobutyl ester

H0110-500ML H0110-25ML
TCH0110-025MLEA 19.29 USD
TCH0110-025ML TCH0110-500ML
Isobutyl hexanoate ≥98.0%
Isobutyl hexanoate

CAS Number: 105-79-3
MDL Number: MFCD00048870
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 76
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Hexanoic Acid Isobutyl Ester

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 348 °C (1013 hPa)
Density: 0.86 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00048870
CAS Number: 105-79-3
EINECS: 203-332-6
https://us.vwr.com/stibo/web/std.lang.all/01/72/28970172.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR