1-Heneicosanol ≥98.0%

Supplier: TCI America

Synonyms: HeneicosanolHeneicosyl alcohol

H0011-10G
TCH0011-010GEA 1182 USD
TCH0011-010G
1-Heneicosanol ≥98.0%
1-Heneicosanol

CAS Number: 15594-90-8
MDL Number: MFCD00062834
Molecular Formula: C21H44O
Molecular Weight: 312.58
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 69


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Heneicosyl Alcohol

Formula: H₃C(CH₂)₁₉CH₂OH
MW: 312.58 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062834
CAS Number: 15594-90-8
EINECS: 239-673-2
https://us.vwr.com/stibo/web/std.lang.all/00/88/28970088.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR