4-Fluorodiphenylmethane ≥95.0%
Supplier: TCI America
F0568-5G
F0568-25G
TCF0568-5GEA
89
USD
TCF0568-5G
TCF0568-25G
4-Fluorodiphenylmethane ≥95.0%
4-Fluorodiphenylmethane
CAS Number: 587-79-1
MDL Number: MFCD00464329
Molecular Formula: C13H11F
Molecular Weight: 186.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 132
Specific Gravity (20/20): 1.09
Formula:
C₁₃H₁₁F Boiling Pt: 132 °C Density: 1.09 (20 °C) |
MDL Number:
MFCD00464329 CAS Number: 587-79-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...