4-Fluoroindole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Fluoro-1H-indole

F0556-1G
TCF0556-1GEA 54 USD
TCF0556-1G
4-Fluoroindole ≥98.0%
4-Fluoroindole

CAS Number: 387-43-9
MDL Number: MFCD00055992
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 90
Melting point (°C): 33

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 90 °C
Melting Pt: 33 °C
MDL Number: MFCD00055992
CAS Number: 387-43-9
https://us.vwr.com/stibo/web/std.lang.all/91/20/28969120.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR