4-Fluorobenzal chloride ≥95.0%
Supplier: TCI America
Synonyms:
α,α-Dichloro-4-fluortoluene
F0315-25G
TCF0315-025GEA
71.03
USD
TCF0315-025G
4-Fluorobenzal chloride ≥95.0%
4-Fluorobenzal chloride
CAS Number: 456-19-9
MDL Number: MFCD00000832
Molecular Formula: C7H5Cl2F
Molecular Weight: 179.02
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 90
Specific Gravity (20/20): 1.35
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Dichloromethyl)-4-fluorobenzene
Formula:
C₇H₅Cl₂F MW: 179.02 g/mol Boiling Pt: 83 °C Density: 1.35 (20 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000832 CAS Number: 456-19-9 EINECS: 207-258-5 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...