Fast black K salt

Supplier: TCI America
Danger

F0092-25G
TCF0092-025GEA 41.67 USD
TCF0092-025G
Fast black K salt
Fast black K salt

CAS Number: 64071-86-9
MDL Number: MFCD00078471
Molecular Formula: C14H12ClN5O4
Molecular Weight: 417.87
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 150
Lambda max.: 455 nm (H2O)


Other Product Names / Synonyms: 2,5-Dimethoxy-4-(4-nitrophenylazo)benzenediazonium Chloride Zinc Chloride

Formula: C₂₈H₂₄Cl₄N₁₀O₈Zn
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00078471
CAS Number: 64071-86-9
EINECS: 264-656-1
https://us.vwr.com/stibo/web/std.lang.all/87/22/28968722.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR