Isobutyl formate ≥95.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Formic acid Isobutyl ester
F0055-500ML
F0055-25ML
TCF0055-025MLEA
30.05
USD
TCF0055-025ML
TCF0055-500ML
Isobutyl formate ≥95.0%
Isobutyl formate
CAS Number: 542-55-2
MDL Number: MFCD00003293
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 10
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Formic Acid Isobutyl Ester
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 98 °C Melting Pt: –96 °C Density: 0.89 (20 °C) Flash Pt: 10 °C |
MDL Number:
MFCD00003293 CAS Number: 542-55-2 EINECS: 208-818-1 UN: 2393 ADR: 3,II Merck Index: 12,05157 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...