1,2-Epoxyhexadecane ≥80.0%

Supplier: TCI America

Synonyms: 1,2-Hexadecylene Oxide

E0316-25ML
TCE0316-025MLEA 42.3 USD
TCE0316-025ML
1,2-Epoxyhexadecane ≥80.0%
1,2-Epoxyhexadecane

CAS Number: 7320-37-8
MDL Number: MFCD00005151
Molecular Formula: C16H32O
Molecular Weight: 240.43
Purity/Analysis Method: 80.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 22
Flash Point (°C): 93


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Hexadecylene Oxide

Formula: C₁₆H₃₂O
MW: 240.43 g/mol
Boiling Pt: 180 °C
Melting Pt: 22 °C
Flash Pt: 93 °C
MDL Number: MFCD00005151
CAS Number: 7320-37-8
EINECS: 230-786-2

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR