2-Ethyl-p-xylene ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Dimethylethylbenzene1-Ethyl-2,5-dimethylbenzene

E0279-5ML
TCE0279-005MLEA 272 USD
TCE0279-005ML
2-Ethyl-p-xylene ≥98.0%
2-Ethyl-p-xylene

CAS Number: 1758-88-9
MDL Number: MFCD00059233
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Melting point (°C): -54
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 2,5-Dimethylethylbenzene, 1-Ethyl-2,5-dimethylbenzene

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 187 °C
Melting Pt: –54 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00059233
CAS Number: 1758-88-9
EINECS: 217-158-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR