2-Ethylnaphthalene ≥99.0%
Supplier: TCI America
E0147-25G
TCE0147-25GEA
210.04
USD
TCE0147-25G
2-Ethylnaphthalene ≥99.0%
2-Ethylnaphthalene
CAS Number: 939-27-5
MDL Number: MFCD00004127
Molecular Formula: C12H12
Molecular Weight: 156.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 252
Melting point (°C): -70
Flash Point (°C): 104
Specific Gravity (20/20): 1.00
Formula:
C₁₂H₁₂ MW: 156.23 g/mol Boiling Pt: 252 °C Melting Pt: –70 °C Density: 1.00 (20 °C) Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004127 CAS Number: 939-27-5 EINECS: 213-360-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...