1-Eicosene ≥85.0%

Supplier: TCI America

Synonyms: 1-Icosene

E0005-25ML
TCE0005-025MLEA 30.45 USD
TCE0005-025ML
1-Eicosene ≥85.0%
1-Eicosene

CAS Number: 3452-07-1
MDL Number: MFCD00009013
Molecular Formula: C20H40
Molecular Weight: 280.54
Purity/Analysis Method: 85.0% (GC)
Form: Crystal
Boiling point (°C): 151
Flash Point (°C): 113
Freezing point (°C): 27


Other Product Names / Synonyms: 1-Icosene

Formula: C₂₀H₄₀
MW: 280.54 g/mol
Boiling Pt: 151 °C
Melting Pt: 60…62 °C
Density: 0.795 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009013
CAS Number: 3452-07-1
EINECS: 222-374-6
https://us.vwr.com/stibo/web/std.lang.all/76/23/28967623.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR