Diallyldiphenylsilane ≥95.0%

Supplier: TCI America

D3730-5G
TCD3730-5GEA 224 USD
TCD3730-5G
Diallyldiphenylsilane ≥95.0%
Diallyldiphenylsilane

CAS Number: 10519-88-7
MDL Number: MFCD00026068
Molecular Formula: C18H20Si
Molecular Weight: 264.44
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 110
Specific Gravity (20/20): 0.99

Formula: C₁₈H₂₀Si
MW: 264.44 g/mol
Boiling Pt: 141 °C
Density: 0.99 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00026068
CAS Number: 10519-88-7
EINECS: 234-061-1
https://us.vwr.com/stibo/web/std.lang.all/57/03/28985703.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR