2,7-Diacetylfluorene ≥98.0%
Supplier: TCI America
D2863-1G
TCD2863-1GEA
35.51
USD
TCD2863-1G
2,7-Diacetylfluorene ≥98.0%
2,7-Diacetylfluorene
CAS Number: 961-27-3
MDL Number: MFCD00045332
Molecular Formula: C17H14O2
Molecular Weight: 250.30
Purity/Analysis Method:
Form: Crystal
Formula:
C₁₇H₁₄O₂ MW: 250.292 g/mol |
MDL Number:
MFCD00045332 CAS Number: 961-27-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...