Diphenyl ditelluride ≥98.0%

Supplier: TCI America

D2718-5G D2718-1G
TCD2718-1GEA 69.02 USD
TCD2718-1G TCD2718-005G
Diphenyl ditelluride ≥98.0%
Diphenyl ditelluride

CAS Number: 32294-60-3
MDL Number: MFCD00192106
Molecular Formula: C12H10Te2
Molecular Weight: 409.41
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 65


Other Product Names / Synonyms: Phenyl Ditelluride

Formula: C₁₂H₁₀Te₂
Melting Pt: 65 °C
MDL Number: MFCD00192106
CAS Number: 32294-60-3
EINECS: 250-982-1
https://us.vwr.com/stibo/web/std.lang.all/98/36/28979836.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR