3,5-Dichloroisonicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,5-Dichloropyridine-4-carboxylic acid

D2600-1G
TCD2600-001GEA 33 USD
TCD2600-001G
3,5-Dichloroisonicotinic acid ≥98.0% (by titrimetric analysis)
3,5-Dichloroisonicotinic acid

CAS Number: 13958-93-5
MDL Number: MFCD01861975
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3,5-Dichloropyridine-4-carboxylic Acid

Formula: C₆H₃Cl₂NO₂
MDL Number: MFCD01861975
CAS Number: 13958-93-5
https://us.vwr.com/stibo/web/std.lang.all/95/69/28979569.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR