2,3,4-Trimethyl-3-pentanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,3,4-Trimethylpentan-3-ol

D2436-25G D2436-5G
TCD2436-005GEA 121 USD
TCD2436-005G TCD2436-25G
2,3,4-Trimethyl-3-pentanol ≥98.0%
2,3,4-Trimethyl-3-pentanol

CAS Number: 3054-92-0
MDL Number: MFCD00048319
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: 1,1-Diisopropylethanol

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 78 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00048319
CAS Number: 3054-92-0
https://us.vwr.com/stibo/web/std.lang.all/93/62/28979362.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR