2,3-Dichlorobenzyl alcohol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
(2,3-Dichlorophenyl)methanol
D2425-5G
TCD2425-005GEA
63.95
USD
TCD2425-005G
2,3-Dichlorobenzyl alcohol ≥98.0%
2,3-Dichlorobenzyl alcohol
CAS Number: 38594-42-2
MDL Number: MFCD00238581
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 86
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD00238581 CAS Number: 38594-42-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...