2,7-Diisopropylnaphthalene ≥97.0%
Supplier: TCI America
D2422-25ML
TCD2422-025MLEA
126.84
USD
TCD2422-025ML
2,7-Diisopropylnaphthalene ≥97.0%
2,7-Diisopropylnaphthalene
CAS Number: 40458-98-8
MDL Number: MFCD00671522
Molecular Formula: C16H20
Molecular Weight: 212.34
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 280
Specific Gravity (20/20): 0.95
Formula:
C₁₆H₂₀ MW: 212.33 g/mol Boiling Pt: 280 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00671522 CAS Number: 40458-98-8 EINECS: 254-929-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...