TCIAD2409

D2409-10G
TCD2409-010GEA 185.28 USD
TCD2409-010G
TCIAD2409
CAS Number: 81029-03-0
MDL Number: MFCD00007163
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 72

Other Product Names / Synonyms: 3-Methoxy-6-nitro-o-xylene, 1-Methoxy-2,3-dimethyl-4-nitrobenzene
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR