4,5-Dichlorophthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione4,5-Dichlorophthalic Anhydride

D2335-5G
TCD2335-005GEA 60 USD
TCD2335-005G
4,5-Dichlorophthalic anhydride ≥98.0% (by GC, titration analysis)
4,5-Dichlorophthalic anhydride

CAS Number: 942-06-3
MDL Number: MFCD00075034
Molecular Formula: C8H2Cl2O3
Molecular Weight: 217.00
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 313
Melting point (°C): 188

Formula: C₈H₂Cl₂O₃
MW: 217.01 g/mol
Boiling Pt: 313 °C
Melting Pt: 188 °C
MDL Number: MFCD00075034
CAS Number: 942-06-3
EINECS: 213-386-2
https://us.vwr.com/stibo/web/std.lang.all/92/68/28979268.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR