Diphenyliodonium hexafluoroarsenate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

D2248-1G
TCD2248-001GEA 108 USD
TCD2248-001G
Diphenyliodonium hexafluoroarsenate ≥98.0% (by titrimetric analysis)
Diphenyliodonium hexafluoroarsenate

CAS Number: 62613-15-4
MDL Number: MFCD00191354
Molecular Formula: C12H10AsF6I
Molecular Weight: 470.03
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 130
Storage Temperature: 0-10°C

Formula: C₁₂H₁₀AsF₆I
MW: 470.03 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00191354
CAS Number: 62613-15-4
EINECS: 263-638-0
https://us.vwr.com/stibo/web/std.lang.all/91/64/28979164.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR