Denatonium benzoate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate
D2124-25G
D2124-5G
D2124-100G
TCD2124-005GEA
91.31
USD
TCD2124-005G
TCD2124-25G
TCD2124-100G
Denatonium benzoate ≥98.0% (by HPLC)
Denatonium benzoate
CAS Number: 3734-33-6
MDL Number: MFCD00031578
Molecular Formula: C28H34N2O3
Molecular Weight: 446.59
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 163
Other Product Names / Synonyms: Benzyldiethyl[(2,6-dimethylphenylcarbamoyl)methyl]ammonium Benzoate
Formula:
C₂₈H₃₄N₂O₃ MW: 446.59 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 EINECS: 223-095-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...