trans-2-Dodecenal ≥90.0%
Supplier: TCI America
|
Danger
|
Synonyms:
(E)-2-Dodecenal
D2068-25ML
TCD2068-025MLEA
43.48
USD
TCD2068-025ML
trans-2-Dodecenal ≥90.0%
trans-2-Dodecenal
CAS Number: 20407-84-5
MDL Number: MFCD00014674
Molecular Formula: C12H22O
Molecular Weight: 182.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 129
Specific Gravity (20/20): 0.85
Formula:
C₁₂H₂₂O MW: 182.31 g/mol Boiling Pt: 132 °C Density: 0.85 (20 °C) Flash Pt: 129 °C |
MDL Number:
MFCD00014674 CAS Number: 20407-84-5 EINECS: 243-797-2 UN: 1760 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...