N,N-Dipropylaniline ≥98.0%

Supplier: TCI America

Synonyms: N-Phenyldipropylamine

D1941-25ML
TCD1941-025MLEA 69 USD
TCD1941-025ML
N,N-Dipropylaniline ≥98.0%
N,N-Dipropylaniline

CAS Number: 2217-07-4
MDL Number: MFCD00035797
Molecular Formula: C12H19N
Molecular Weight: 177.29
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 92
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N-Phenyldipropylamine

Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 112 °C
Density: 0.92 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00035797
CAS Number: 2217-07-4
EINECS: 218-705-9
https://us.vwr.com/stibo/web/std.lang.all/87/42/28978742.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR