N,N-Dimethylisopropylamine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-Isopropyldimethylamine
D1563-25ML
D1563-500ML
TCD1563-025MLEA
22.31
USD
TCD1563-025ML
TCD1563-500ML
N,N-Dimethylisopropylamine ≥99.0% (by GC, titration analysis)
N,N-Dimethylisopropylamine
CAS Number: 996-35-0
MDL Number: MFCD00015014
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 68
Flash Point (°C): -27
Specific Gravity (20/20): 0.72
Other Product Names / Synonyms: N-Isopropyldimethylamine
Formula:
C₅H₁₃N MW: 87.16 g/mol Boiling Pt: 68 °C Density: 0.72 (20 °C) Flash Pt: –27 °C |
MDL Number:
MFCD00015014 CAS Number: 996-35-0 EINECS: 213-635-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...