2,4-Dimethyl-2-pentene ≥98.0%

Supplier: TCI America

D1254-5ML
TCD1254-005MLEA 439 USD
TCD1254-005ML
2,4-Dimethyl-2-pentene ≥98.0%
2,4-Dimethyl-2-pentene

CAS Number: 625-65-0
MDL Number: MFCD00026435
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Melting point (°C): -128
Specific Gravity (20/20): 0.70
Storage Temperature: 0-10°C

Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 83 °C
Melting Pt: –128 °C
Density: 0.70 (20 °C)
MDL Number: MFCD00026435
CAS Number: 625-65-0
EINECS: 210-905-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR