3,6-Dimethylphenanthrene ≥97.0%
Supplier: TCI America
D1235-100MG
TCD1235-100MGEA
451.48
USD
TCD1235-100MG
3,6-Dimethylphenanthrene ≥97.0%
3,6-Dimethylphenanthrene
CAS Number: 1576-67-6
MDL Number: MFCD00001170
Molecular Formula: C16H14
Molecular Weight: 206.29
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 141
Formula:
C₁₆H₁₄ MW: 206.29 g/mol Melting Pt: 141 °C |
MDL Number:
MFCD00001170 CAS Number: 1576-67-6 EINECS: 216-409-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...