Dodecylcyclohexane ≥98.0%
Supplier: TCI America
Synonyms:
1-Cyclohexyldodecane
, Laurylcyclohexane
D1232-25ML
D1232-500ML
TCD1232-025MLEA
144.07
USD
TCD1232-025ML
TCD1232-500ML
Dodecylcyclohexane ≥98.0%
Dodecylcyclohexane
CAS Number: 1795-17-1
MDL Number: MFCD00045506
Molecular Formula: C18H36
Molecular Weight: 252.49
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: 1-Cyclohexyldodecane, Laurylcyclohexane
Formula:
C₁₈H₃₆ MW: 252.48 g/mol Boiling Pt: 126 °C Melting Pt: 12.5 °C Density: 0.82 (20 °C) |
MDL Number:
MFCD00045506 CAS Number: 1795-17-1 EINECS: 217-273-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...