2,3-Dimethylheptane ≥99.0%

Supplier: TCI America

D1203-1ML
TCD1203-001MLEA 170.31 USD
TCD1203-001ML
2,3-Dimethylheptane ≥99.0%
2,3-Dimethylheptane

CAS Number: 3074-71-3
MDL Number: MFCD00048819
Molecular Formula: C9H20
Molecular Weight: 128.26
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 26
Specific Gravity (20/20): 0.73

Formula: C₉H₂₀
MW: 128.26 g/mol
Boiling Pt: 140 °C
Melting Pt: –112 °C
Density: 0.73 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00048819
CAS Number: 3074-71-3

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR