5-Dodecyne ≥98.0%
Supplier: TCI America
D1191-5ML
TCD1191-005MLEA
238
USD
TCD1191-005ML
5-Dodecyne ≥98.0%
5-Dodecyne
CAS Number: 19780-12-2
MDL Number: MFCD00041661
Molecular Formula: C12H22
Molecular Weight: 166.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 93
Specific Gravity (20/20): 0.78
Formula:
C₁₂H₂₂ MW: 166.31 g/mol Boiling Pt: 93 °C Density: 0.78 (20 °C) |
MDL Number:
MFCD00041661 CAS Number: 19780-12-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...