1,1-Diphenylhydrazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D1161-5ML
TCD1161-005MLEA 173 USD
TCD1161-005ML
1,1-Diphenylhydrazine ≥97.0% (by GC, titration analysis)
1,1-Diphenylhydrazine

CAS Number: 530-50-7
MDL Number: MFCD00044505
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 35

Formula: C₁₂H₁₂N₂
Boiling Pt: 220 °C
Melting Pt: 35 °C
MDL Number: MFCD00044505
CAS Number: 530-50-7
EINECS: 208-483-1

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR