2,4-Dimethylhexane ≥98.0%

Supplier: TCI America
Danger

D1110-5ML
TCD1110-005MLEA 50.73 USD
TCD1110-005ML
2,4-Dimethylhexane ≥98.0%
2,4-Dimethylhexane

CAS Number: 589-43-5
MDL Number: MFCD00039929
Molecular Formula: C8H18
Molecular Weight: 114.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 10
Specific Gravity (20/20): 0.70

Formula: C₈H₁₈
MW: 114.23 g/mol
Boiling Pt: 109 °C
Melting Pt: –124 °C
Density: 0.70 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00039929
CAS Number: 589-43-5
EINECS: 209-649-6
UN: 1262
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR