3,4-Dichloro-1-butene ≥98.0%

Supplier: TCI America

D1072-25G
TCD1072-025GEA 117.72 USD
TCD1072-025G
3,4-Dichloro-1-butene ≥98.0%
3,4-Dichloro-1-butene

CAS Number: 760-23-6
MDL Number: MFCD00000947
Molecular Formula: C4H6Cl2
Molecular Weight: 124.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Melting point (°C): -61
Flash Point (°C): 28
Specific Gravity (20/20): 1.16

Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 122 °C
Melting Pt: –61 °C
Density: 1.16 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000947
CAS Number: 760-23-6
EINECS: 212-079-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR