1-Docosene ≥99.0%

Supplier: TCI America

D1013-25G
TCD1013-025GEA 180.92 USD
TCD1013-025G
1-Docosene ≥99.0%
1-Docosene

CAS Number: 1599-67-3
MDL Number: MFCD00027085
Molecular Formula: C22H44
Molecular Weight: 308.59
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Freezing point (°C): 38

Formula: C₂₂H₄₄
MW: 308.59 g/mol
Boiling Pt: 162 °C (0.1 mmHg)
Melting Pt: 58…60 °C
Density: 0.8002 g/cm³ (20 °C)
MDL Number: MFCD00027085
CAS Number: 1599-67-3
EINECS: 216-490-6
https://us.vwr.com/stibo/web/std.lang.all/62/03/28976203.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR