Diphenylamine hydrochloride ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America

D0874-25G
TCD0874-025GEA 52.75 USD
TCD0874-025G
Diphenylamine hydrochloride ≥99.0% (by HPLC, total nitrogen)
Diphenylamine hydrochloride

CAS Number: 537-67-7
MDL Number: MFCD00042007
Molecular Formula: C12H11N
Molecular Weight: 205.69
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 180
Storage Temperature: 0°C

Formula: C₁₂H₁₂ClN
MW: 205.69 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00042007
CAS Number: 537-67-7
EINECS: 208-675-5
https://us.vwr.com/stibo/web/std.lang.all/59/82/28975982.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR