N,N-Diphenylacetamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: N-Acetyldiphenylamine

D0868-25G
TCD0868-025GEA 77.14 USD
TCD0868-025G
N,N-Diphenylacetamide ≥98.0% (by total nitrogen basis)
N,N-Diphenylacetamide

CAS Number: 519-87-9
MDL Number: MFCD00008685
Molecular Formula: C14H13NO
Molecular Weight: 211.26
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: N-Acetyldiphenylamine

Formula: C₁₄H₁₃NO
Melting Pt: 100 °C
MDL Number: MFCD00008685
CAS Number: 519-87-9
EINECS: 208-277-1
https://us.vwr.com/stibo/web/std.lang.all/59/71/28975971.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR