cis-1,2-Dimethylcyclohexane ≥98.0%

Supplier: TCI America

D0696-25ML
TCD0696-025MLEA 96.39 USD
TCD0696-025ML
cis-1,2-Dimethylcyclohexane ≥98.0%
cis-1,2-Dimethylcyclohexane

CAS Number: 2207-01-4
MDL Number: MFCD00063621
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 12
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: cis-Hexahydro-o-xylene

Formula: C₈H₁₆
Boiling Pt: 130 °C
Melting Pt: –90 °C
Density: 0.80 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00063621
CAS Number: 2207-01-4
EINECS: 218-621-2
UN: 2263
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/56/17/28975617.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR