N,N-Diethyl-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N,N-Diethyl-4-methylaniline
, 4-Diethylaminotoluene
D0533-25ML
TCD0533-025MLEA
99.44
USD
TCD0533-025ML
N,N-Diethyl-p-toluidine ≥98.0% (by GC, titration analysis)
N,N-Diethyl-p-toluidine
CAS Number: 613-48-9
MDL Number: MFCD00015113
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 102
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 4-Diethylaminotoluene, N,N-Diethyl-4-methylaniline
Formula:
C₁₁H₁₇N MW: 163.26 g/mol Boiling Pt: 230 °C Density: 0.92 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00015113 CAS Number: 613-48-9 EINECS: 210-345-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...