1,3-Dichlorobutane ≥98.0%
Supplier: TCI America
|
Danger
|
D0348-25G
TCD0348-025GEA
103.53
USD
TCD0348-025G
1,3-Dichlorobutane ≥98.0%
1,3-Dichlorobutane
CAS Number: 1190-22-3
MDL Number: MFCD00000870
Molecular Formula: C4H8Cl2
Molecular Weight: 127.01
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 31
Specific Gravity (20/20): 1.12
Formula:
ClH₂CCH₂CHClCH₃ MW: 127.01 g/mol Boiling Pt: 134 °C Density: 1.12 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00000870 CAS Number: 1190-22-3 EINECS: 214-718-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...