1,10-Diaminodecane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
1,10-Decanediamine
D0085-25G
TCD0085-025GEA
139
USD
TCD0085-025G
1,10-Diaminodecane ≥98.0% (by GC, titration analysis)
1,10-Diaminodecane
CAS Number: 646-25-3
MDL Number: MFCD00008151
Molecular Formula: C10H24N2
Molecular Weight: 172.32
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 61
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Decamethylenediamine, 1,10-Decanediamine
Formula:
C₁₀H₂₄N₂ MW: 172.31 g/mol Boiling Pt: 140 °C Melting Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008151 CAS Number: 646-25-3 EINECS: 211-471-9 UN: 3259 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...