cis-Decahydronaphthalene ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
cis-Decalin
, cis-Bicyclo[4.4.0]decane
D0009-500ML
D0009-25ML
TCD0009-025MLEA
65.96
USD
TCD0009-025ML
TCD0009-500ML
cis-Decahydronaphthalene ≥98.0%
cis-Decahydronaphthalene
CAS Number: 493-01-6
MDL Number: MFCD00064189
Molecular Formula: C10H18
Molecular Weight: 138.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 58
Specific Gravity (20/20): 0.90
Formula:
C₁₀H₁₈ MW: 138.25 g/mol Boiling Pt: 193 °C Melting Pt: –43 °C Density: 0.90 (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064189 CAS Number: 493-01-6 EINECS: 207-770-9 UN: 1147 ADR: 3,III Merck Index: 13,02866 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...