5-Chloro-2-iodoaniline ≥95.0% (by GC, titration analysis)
Supplier: TCI America
C2239-5G
TCC2239-5GEA
40.57
USD
TCC2239-5G
5-Chloro-2-iodoaniline ≥95.0% (by GC, titration analysis)
5-Chloro-2-iodoaniline
CAS Number: 6828-35-9
MDL Number: MFCD06738960
Molecular Formula: C6H5ClIN
Molecular Weight: 253.47
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 42
Formula:
C₆H₅ClIN MW: 253.47 g/mol Melting Pt: 42 °C |
MDL Number:
MFCD06738960 CAS Number: 6828-35-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...