2-Chloro-5-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2077-25G C2077-5G
TCC2077-25GEA 141.09 USD
TCC2077-25G TCC2077-5G
2-Chloro-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-5-nitrophenol

CAS Number: 619-10-3
Molecular Formula: C6H4ClNO3
Molecular Weight: 173.55
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 112

Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 112 °C
MDL Number: MFCD01571825
CAS Number: 619-10-3
EINECS: 210-579-3
https://us.vwr.com/stibo/web/std.lang.all/91/38/28939138.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR