2-Chlorocycloheptanone ≥98.0%

C1645-5G
TCC1645-5GEA 316.21 USD
TCC1645-5G
2-Chlorocycloheptanone ≥98.0%
2-Chlorocycloheptanone

CAS Number: 766-66-5
MDL Number: MFCD00041033
Molecular Formula: C7H11ClO
Molecular Weight: 146.61
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.14
Storage Temperature: 0°C

Formula: C₇H₁₁ClO
MW: 146.62 g/mol
Density: 1.14 (20 °C)
MDL Number: MFCD00041033
CAS Number: 766-66-5
EINECS: 212-168-4
https://us.vwr.com/stibo/web/std.lang.all/88/60/28938860.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR